Product Details
- SNP ID
-
rs147584630
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:900542 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGCCGCCACTCACATCTGCGTC[A/G]CCGGCCCCTGGGAATGCCACCTGCA
- Phenotype
-
MIM: 608114
MIM: 612056
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10256887] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAP1
- Gene Name
- ArfGAP with dual PH domains 1
- Gene
- GET4
- Gene Name
- golgi to ER traffic protein 4
There are no transcripts associated with this gene.
View Full Product Details