Product Details

SNP ID
rs147665999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30014859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTTTCATTCACCACCGCACCA[G/T]GTTTTTCAAACTCCTTCTTTAAATA
Phenotype
MIM: 614505
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FKBP14 PubMed Links

Gene Details

Gene
FKBP14
Gene Name
FK506 binding protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017946.3 805 Missense Mutation CAT,CCT H171P NP_060416.1

View Full Product Details