Product Details

SNP ID
rs147996458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44115799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGGCTGTGGGGCTGATGTGAC[A/G]GACCCGCAGGGTCCACTCCAGGATC
Phenotype
MIM: 602981 MIM: 600815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AEBP1 PubMed Links

Gene Details

Gene
AEBP1
Gene Name
AE binding protein 1
There are no transcripts associated with this gene.

Gene
MIR4649
Gene Name
microRNA 4649
There are no transcripts associated with this gene.

Gene
POLD2
Gene Name
DAN polymerase delta 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127218.2 1764 Missense Mutation CGT,TGT R372C NP_001120690.1
NM_001256879.1 1764 Missense Mutation CGT,TGT R372C NP_001243808.1
NM_006230.3 1764 Missense Mutation CGT,TGT R407C NP_006221.2
XM_011515431.1 1764 Missense Mutation CGT,TGT R405C XP_011513733.1

View Full Product Details