Product Details
- SNP ID
-
rs148444002
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73769390 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGTCTCCCTGCGTCTGTCCCTTA[A/G]ACGTAGTCCTTGCGGTCGTAGCCTG
- Phenotype
-
MIM: 602910
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN3
PubMed Links
Gene Details
- Gene
- CLDN3
- Gene Name
- claudin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306.3 |
881 |
Silent Mutation |
GTC,GTT |
V220V |
NP_001297.1 |
View Full Product Details