Product Details

SNP ID
rs148507237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:25226699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGTAAAATAAGTCATTGGCAC[A/G]TGGTGAATGCATGGATCCTTGACAC
Phenotype
MIM: 616984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPVF PubMed Links

Gene Details

Gene
NPVF
Gene Name
neuropeptide VF precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022150.3 513 Missense Mutation NP_071433.3

View Full Product Details