Product Details

SNP ID
rs148977943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19722090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACATTGATTACACTCTTCCATT[A/G]AATATCAGCTGTGGTCCTCCATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM196 PubMed Links

Gene Details

Gene
TMEM196
Gene Name
transmembrane protein 196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152774.3 1526 Silent Mutation TTC,TTT F168F NP_689987.3
XM_005249692.4 1526 UTR 3 XP_005249749.2
XM_011515249.2 1526 UTR 3 XP_011513551.1
XM_011515250.2 1526 Silent Mutation TTC,TTT F174F XP_011513552.1
XM_017011928.1 1526 Intron XP_016867417.1
XM_017011929.1 1526 Intron XP_016867418.1

View Full Product Details