Product Details

SNP ID
rs148980420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92199373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGTAATATAAAAATTATCTGC[A/C]AACTGTAGGACAACTTTAAATATGA
Phenotype
MIM: 604214
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRIT1 PubMed Links

Gene Details

Gene
KRIT1
Gene Name
KRIT1, ankyrin repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013406.1 4593 UTR 3 NP_001013424.1
NM_004912.3 4593 UTR 3 NP_004903.2
NM_194454.1 4593 UTR 3 NP_919436.1
NM_194455.1 4593 UTR 3 NP_919437.1
NM_194456.1 4593 UTR 3 NP_919438.1
XM_005250660.3 4593 Intron XP_005250717.1
XM_005250662.3 4593 Intron XP_005250719.1
XM_005250665.3 4593 Intron XP_005250722.1
XM_005250666.3 4593 Intron XP_005250723.1
XM_005250667.2 4593 Intron XP_005250724.1
XM_005250668.3 4593 Intron XP_005250725.1
XM_005250669.3 4593 Intron XP_005250726.1
XM_006716161.3 4593 Intron XP_006716224.1
XM_006716162.3 4593 Intron XP_006716225.1
XM_006716163.3 4593 Intron XP_006716226.1
XM_011516651.2 4593 Intron XP_011514953.1
XM_011516653.2 4593 Intron XP_011514955.1
XM_011516654.2 4593 Intron XP_011514956.1
XM_011516655.2 4593 Intron XP_011514957.1
XM_011516656.2 4593 Intron XP_011514958.1
XM_011516657.2 4593 Intron XP_011514959.1
XM_011516658.2 4593 Intron XP_011514960.1
XM_011516659.2 4593 Intron XP_011514961.1
XM_011516660.2 4593 Intron XP_011514962.1
XM_011516661.2 4593 Intron XP_011514963.1
XM_017012755.1 4593 Intron XP_016868244.1
XM_017012756.1 4593 Intron XP_016868245.1
XM_017012757.1 4593 Intron XP_016868246.1
XM_017012758.1 4593 Intron XP_016868247.1
XM_017012759.1 4593 Intron XP_016868248.1
Gene
MIR1285-1
Gene Name
microRNA 1285-1
There are no transcripts associated with this gene.

View Full Product Details