Product Details

SNP ID
rs149479978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149842132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGAAATCCTTCCTGAAAGAGAA[C/T]GTGAATCGCGACTTTATGGGTGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSPO PubMed Links

Gene Details

Gene
SSPO
Gene Name
SCO-spondin
There are no transcripts associated with this gene.

Gene
ZNF862
Gene Name
zinc finger protein 862
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099220.1 Intron NP_001092690.1
XM_006716085.3 Intron XP_006716148.1
XM_011516463.1 Intron XP_011514765.1
XM_011516464.1 Intron XP_011514766.1
XM_011516465.2 Intron XP_011514767.1
XM_011516466.2 Intron XP_011514768.1
XM_017012519.1 Intron XP_016868008.1

View Full Product Details