Product Details

SNP ID
rs149615400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142941317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGTGCACAGCGGAAATACCTGG[C/T]AAAGGCTGGGGTGCTGCTGAGGGGC
Phenotype
MIM: 613883 MIM: 606679
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf34 PubMed Links

Gene Details

Gene
C7orf34
Gene Name
chromosome 7 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178829.4 1515 Intron NP_849151.2
Gene
KEL
Gene Name
Kell blood group, metallo-endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000420.2 1515 Missense Mutation ACC,GCC T712A NP_000411.1
XM_005249993.2 1515 Missense Mutation ACC,GCC T724A XP_005250050.1
XM_005249994.3 1515 Missense Mutation ACC,GCC T403A XP_005250051.1
Gene
TRPV5
Gene Name
transient receptor potential cation channel subfamily V member 5
There are no transcripts associated with this gene.

View Full Product Details