Product Details

SNP ID
rs149861210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:25136324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGGTCTGGACACATATTATAC[G/T]GATCACGTAGGGGAGATTTAAGATT
Phenotype
MIM: 616071
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C7orf31 PubMed Links

Gene Details

Gene
C7orf31
Gene Name
chromosome 7 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138811.3 873 Missense Mutation CAG,CCG Q474P NP_620166.3
XM_005249619.3 873 Missense Mutation CAG,CCG Q474P XP_005249676.1
XM_005249620.3 873 Missense Mutation CAG,CCG Q474P XP_005249677.1
XM_011515125.2 873 Missense Mutation CAG,CCG Q264P XP_011513427.1

View Full Product Details