Product Details

SNP ID
rs150138159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141778952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATCACGCTTAGCCAGGCATCAC[C/T]TTTTCCTGAACTTGTGACTACGAGA
Phenotype
MIM: 604869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R4 PubMed Links

Gene Details

Gene
TAS2R4
Gene Name
taste 2 receptor member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016944.1 464 Missense Mutation CCT,CTT P155L NP_058640.1

View Full Product Details