Product Details

SNP ID
rs150285912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:83961449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTATTTTCTTGTAAGTGCTTCCA[C/T]TTGTTACTGTTCCCTGGGGTATGTC
Phenotype
MIM: 603961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA3A PubMed Links

Gene Details

Gene
SEMA3A
Gene Name
semaphorin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006080.2 2588 Silent Mutation AAA,AAG K746K NP_006071.1
XM_005250110.3 2588 Silent Mutation AAA,AAG K746K XP_005250167.1
XM_005250111.4 2588 Silent Mutation AAA,AAG K746K XP_005250168.1
XM_006715839.3 2588 Silent Mutation AAA,AAG K746K XP_006715902.1
XM_011515734.2 2588 Silent Mutation AAA,AAG K746K XP_011514036.1
XM_017011673.1 2588 Silent Mutation AAA,AAG K746K XP_016867162.1

View Full Product Details