Product Details

SNP ID
rs151212554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108021281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAAGTGGACCTCAGCTGGAGG[C/T]CAAGGTTTCAGAGCTGGATGCACTT
Phenotype
MIM: 616380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMB4 PubMed Links

Gene Details

Gene
LAMB4
Gene Name
laminin subunit beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318046.1 Intron NP_001304975.1
NM_001318047.1 Intron NP_001304976.1
NM_001318048.1 Intron NP_001304977.1
NM_007356.2 Intron NP_031382.2
XM_011515975.1 Intron XP_011514277.1
XM_011515978.1 Intron XP_011514280.1
XM_011515979.2 Intron XP_011514281.1
XM_011515980.2 Intron XP_011514282.1
XM_017011879.1 Intron XP_016867368.1
XM_017011880.1 Intron XP_016867369.1

View Full Product Details