Product Details

SNP ID
rs111321152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52940009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGACTCTGGCGCCGCTGCCGGC[A/G]CCGCTGGCGGTGGCTGTACCAGTTG
Phenotype
MIM: 600730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPBWR1 PubMed Links

Gene Details

Gene
NPBWR1
Gene Name
neuropeptides B/W receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005285.3 102 Silent Mutation GCA,GCG A34A NP_005276.2

View Full Product Details