Product Details

SNP ID
rs111439067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41264863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCCCCTTTTTGTGTTTTAGTGG[C/T]TGTGTTGCTGTTGGAAATGGTTTGT
Phenotype
MIM: 604156
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR548AO PubMed Links

Gene Details

Gene
MIR548AO
Gene Name
microRNA 548ao
There are no transcripts associated with this gene.

Gene
SFRP1
Gene Name
secreted frizzled related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003012.4 1561 UTR 3 NP_003003.3

View Full Product Details