Product Details

SNP ID
rs111478144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66719744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTATTTCTGTTGCTGACATA[C/T]ATACCATTTTGAGTCTTTTAAAATA
Phenotype
MIM: 607591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf44-SGK3 PubMed Links

Gene Details

Gene
C8orf44-SGK3
Gene Name
C8orf44-SGK3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204173.1 Intron NP_001191102.1
Gene
SGK3
Gene Name
serum/glucocorticoid regulated kinase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033578.2 Intron NP_001028750.1
NM_013257.4 Intron NP_037389.4
NM_170709.2 Intron NP_733827.2

View Full Product Details