Product Details

SNP ID
rs112536089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85465400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTCTGTTTCCTATGATCAAGCA[A/T]CTTCCCTGAGGATCCTCAACAATGG
Phenotype
MIM: 611492
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CA2 PubMed Links

Gene Details

Gene
CA2
Gene Name
carbonic anhydrase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000067.2 343 Missense Mutation ACT,TCT T55S NP_000058.1
NM_001293675.1 343 UTR 5 NP_001280604.1
Gene
CA3-AS1
Gene Name
CA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details