Product Details

SNP ID
rs113170706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39936978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGAAAGACATCTTTTAAAAGA[A/G]GAATAATCATGACTAGTTTAATCAT
Phenotype
MIM: 147435 MIM: 612129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDO1 PubMed Links

Gene Details

Gene
IDO1
Gene Name
indoleamine 2,3-dioxygenase 1
There are no transcripts associated with this gene.

Gene
IDO2
Gene Name
indoleamine 2,3-dioxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194294.2 Intron NP_919270.2

View Full Product Details