Product Details

SNP ID
rs113782797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30794016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAAATGTGTAGCCAGCACCACG[C/T]GGTGAAATACCCCATCCACCACGAT
Phenotype
MIM: 176916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP2CB PubMed Links

Gene Details

Gene
PPP2CB
Gene Name
protein phosphatase 2 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009552.1 1054 Silent Mutation CCA,CCG P213P NP_001009552.1

View Full Product Details