Product Details

SNP ID
rs114449615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73066566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATTATTTTATTCAGCTTCAGGC[C/T]TTAAGCCTTGGAAATAATCTTAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SBSPON PubMed Links

Gene Details

Gene
SBSPON
Gene Name
somatomedin B and thrombospondin type 1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153225.3 1775 UTR 3 NP_694957.3
XM_017013145.1 1775 UTR 3 XP_016868634.1

View Full Product Details