Product Details

SNP ID
rs116651927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:63060220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAAGTATGATTCCATCCCTTTC[C/T]GTGTAGTCATAAGAATGACCATAAG
Phenotype
MIM: 600415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTPA PubMed Links

Gene Details

Gene
TTPA
Gene Name
alpha tocopherol transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000370.3 2023 UTR 3 NP_000361.1
XM_006716468.3 2023 UTR 3 XP_006716531.1

View Full Product Details