Product Details

SNP ID
rs116877116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:134478098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAGCGGTTGCAGATGAACATTT[A/G]GCACCTAGATGGGGGTCAAGGAGCT
Phenotype
MIM: 610931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFAT PubMed Links

Gene Details

Gene
ZFAT
Gene Name
zinc finger and AT-hook domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029939.3 4461 UTR 3 NP_001025110.2
NM_001167583.2 4461 UTR 3 NP_001161055.1
NM_001174157.1 4461 UTR 3 NP_001167628.1
NM_001174158.1 4461 UTR 3 NP_001167629.1
NM_001289394.1 4461 UTR 3 NP_001276323.1
NM_020863.3 4461 UTR 3 NP_065914.2
XM_011517203.1 4461 UTR 3 XP_011515505.1
XM_011517204.2 4461 UTR 3 XP_011515506.1
XM_011517206.1 4461 Intron XP_011515508.1
XM_017013716.1 4461 Intron XP_016869205.1

View Full Product Details