Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001291931.1 | 2227 | Intron | NP_001278860.1 | ||
NM_145003.4 | 2227 | UTR 3 | NP_659440.2 | ||
XM_011516912.1 | 2227 | Intron | XP_011515214.1 | ||
XM_011516913.2 | 2227 | Intron | XP_011515215.1 | ||
XM_011516914.1 | 2227 | Intron | XP_011515216.1 | ||
XM_011516915.2 | 2227 | Intron | XP_011515217.1 | ||
XM_011516916.1 | 2227 | Intron | XP_011515218.1 | ||
XM_011516917.2 | 2227 | Intron | XP_011515219.1 | ||
XM_011516918.1 | 2227 | Intron | XP_011515220.1 | ||
XM_011516919.1 | 2227 | Intron | XP_011515221.1 | ||
XM_011516920.1 | 2227 | Intron | XP_011515222.1 | ||
XM_011516921.1 | 2227 | Intron | XP_011515223.1 | ||
XM_011516922.1 | 2227 | Intron | XP_011515224.1 | ||
XM_011516923.2 | 2227 | Intron | XP_011515225.1 | ||
XM_011516924.1 | 2227 | Intron | XP_011515226.1 | ||
XM_017013175.1 | 2227 | Intron | XP_016868664.1 | ||
XM_017013176.1 | 2227 | UTR 3 | XP_016868665.1 | ||
XM_017013177.1 | 2227 | UTR 3 | XP_016868666.1 | ||
XM_017013178.1 | 2227 | UTR 3 | XP_016868667.1 | ||
XM_017013179.1 | 2227 | Intron | XP_016868668.1 | ||
XM_017013180.1 | 2227 | Intron | XP_016868669.1 | ||
XM_017013181.1 | 2227 | Intron | XP_016868670.1 |