Product Details

SNP ID
rs117806436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30390477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAATCCTTCAGTGCTACTTGGTAC[C/T]CCACAAAGTCAAGTGCGCACATGGC
Phenotype
MIM: 601558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBPMS PubMed Links

Gene Details

Gene
RBPMS
Gene Name
RNA binding protein with multiple splicing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008710.2 Intron NP_001008710.1
NM_001008711.2 Intron NP_001008711.1
NM_001008712.2 Intron NP_001008712.1
NM_006867.3 Intron NP_006858.1
XM_017012980.1 Intron XP_016868469.1
XM_017012981.1 Intron XP_016868470.1
XM_017012982.1 Intron XP_016868471.1
XM_017012983.1 Intron XP_016868472.1
XM_017012984.1 Intron XP_016868473.1
XM_017012985.1 Intron XP_016868474.1
XM_017012986.1 Intron XP_016868475.1
XM_017012987.1 Intron XP_016868476.1
XM_017012988.1 Intron XP_016868477.1
XM_017012989.1 Intron XP_016868478.1
XM_017012990.1 Intron XP_016868479.1
XM_017012991.1 Intron XP_016868480.1
XM_017012992.1 Intron XP_016868481.1
Gene
RBPMS-AS1
Gene Name
RBPMS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details