Product Details
- SNP ID
-
rs118007354
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10897815 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTAATTCTTTTTTTTTTGTAGT[A/G]GTGGTGTTGGTGTGGCGGTGTTGGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XKR6
PubMed Links
Gene Details
- Gene
- XKR6
- Gene Name
- XK related 6
View Full Product Details