Product Details

SNP ID
rs118033949
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58565860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTAGAATTTCTTCAGCCGTTGT[C/T]TATTGTTTTAATTTGACAAGATAAA
Phenotype
MIM: 602217
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SDCBP PubMed Links

Gene Details

Gene
SDCBP
Gene Name
syndecan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007067.1 Intron NP_001007068.1
NM_001007068.1 Intron NP_001007069.1
NM_001007069.1 Intron NP_001007070.1
NM_001007070.1 Intron NP_001007071.1
NM_005625.3 Intron NP_005616.2
XM_011517571.2 Intron XP_011515873.2
XM_017013727.1 Intron XP_016869216.1
XM_017013728.1 Intron XP_016869217.1

View Full Product Details