Product Details
- SNP ID
-
rs138261253
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:120451198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTAGTTCTGATTGGCAAGAGATA[C/G]ATTTTGATACAGAAAAAGATAAAAT
- Phenotype
-
MIM: 610200
MIM: 605927
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61753755] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL13
- Gene Name
- mitochondrial ribosomal protein L13
There are no transcripts associated with this gene.
- Gene
- MTBP
- Gene Name
- MDM2 binding protein
View Full Product Details