Product Details

SNP ID
rs138368316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42552656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAATTGTTTCAAGCTCCTGATT[C/G]TTTCTACTAAATCATGAACAGCTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMIM19 PubMed Links

Gene Details

Gene
SMIM19
Gene Name
small integral membrane protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135674.1 1128 UTR 3 NP_001129146.1
NM_001135675.1 1128 UTR 3 NP_001129147.1
NM_001135676.1 1128 UTR 3 NP_001129148.1
NM_138436.3 1128 UTR 3 NP_612445.2
XM_005273398.4 1128 UTR 3 XP_005273455.1

View Full Product Details