Product Details

SNP ID
rs138800445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144317342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGGGACACTCACCACCAGTCC[C/T]GGTAGAACTCCCGGTCTCCAAACTG
Phenotype
MIM: 604900 MIM: 140580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGAT1 PubMed Links

Gene Details

Gene
DGAT1
Gene Name
diacylglycerol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012079.5 1089 Missense Mutation CAG,CGG Q362R NP_036211.2
XM_011517356.2 1089 Missense Mutation CAG,CGG Q305R XP_011515658.1
Gene
HSF1
Gene Name
heat shock transcription factor 1
There are no transcripts associated with this gene.

Gene
MIR6848
Gene Name
microRNA 6848
There are no transcripts associated with this gene.

View Full Product Details