Product Details

SNP ID
rs140277404
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:48723601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCCACTGCTTACTGTGTGTAC[C/T]AAGAAGGCAGAAAGCAGCTCACCCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EFCAB1 PubMed Links

Gene Details

Gene
EFCAB1
Gene Name
EF-hand calcium binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142857.1 2040 Intron NP_001136329.1
NM_024593.3 2040 UTR 3 NP_078869.1
XM_005251303.1 2040 UTR 3 XP_005251360.1
XM_011517589.1 2040 Intron XP_011515891.1
XM_011517590.2 2040 UTR 3 XP_011515892.1
XM_011517591.2 2040 UTR 3 XP_011515893.1
XM_017013826.1 2040 UTR 3 XP_016869315.1
XM_017013827.1 2040 UTR 3 XP_016869316.1
XM_017013828.1 2040 Intron XP_016869317.1

View Full Product Details