Product Details
- SNP ID
-
hCV173952859
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144383123 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGTGGAGGAAGGTGCTCTCCAC[C/T]GCGGTAGTGGGGGCGCCCCTGCTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADCK5
PubMed Links
Gene Details
- Gene
- ADCK5
- Gene Name
- aarF domain containing kinase 5
There are no transcripts associated with this gene.
View Full Product Details