Product Details

SNP ID
rs141054393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100704328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGGAACACCGGTGGCACTGTT[A/T]ACTGCTTTCTGGGCAGCCTCTTTAG
Phenotype
MIM: 604679
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR7705 PubMed Links

Gene Details

Gene
MIR7705
Gene Name
microRNA 7705
There are no transcripts associated with this gene.

Gene
PABPC1
Gene Name
poly(A) binding protein cytoplasmic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002568.3 3296 Intron NP_002559.2
XM_005250861.2 3296 Silent Mutation GTA,GTT V627V XP_005250918.1

View Full Product Details