Product Details

SNP ID
rs141179680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142911989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCAGATGCAAGACTAGTTAAT[C/T]GCTCTGAAAGTGAGGAGGGGGGACG
Phenotype
MIM: 124080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP11B2 PubMed Links

Gene Details

Gene
CYP11B2
Gene Name
cytochrome P450 family 11 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000498.3 1506 Silent Mutation GCA,GCG A501A NP_000489.3

View Full Product Details