Product Details

SNP ID
rs141190521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123681291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGAAGTCCCCGGAGGTATCTGA[A/G]CGAACCATGTCAGAGAGAGACTTCT
Phenotype
MIM: 602573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA13 PubMed Links

Gene Details

Gene
ANXA13
Gene Name
annexin A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003954.2 992 Silent Mutation CGC,CGT R341R NP_001003954.1
NM_004306.3 992 Silent Mutation CGC,CGT R300R NP_004297.2

View Full Product Details