Product Details

SNP ID
rs141648126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:29721672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAGAGCCAGAGATCATGTGTCA[A/G]CACCCAGAGCGGAGGAGAAAGAATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00589 PubMed Links

Gene Details

Gene
LINC00589
Gene Name
long intergenic non-protein coding RNA 589
There are no transcripts associated with this gene.

View Full Product Details