Product Details
- SNP ID
-
rs141909863
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144360149 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAGGGTCTTCTGCTGCTGTTGCC[A/G]CCACCACCATCTGTACCCACAGGGG
- Phenotype
-
MIM: 609076
MIM: 607882
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXL6
PubMed Links
Gene Details
- Gene
- FBXL6
- Gene Name
- F-box and leucine rich repeat protein 6
There are no transcripts associated with this gene.
- Gene
- LOC101928902
- Gene Name
- uncharacterized LOC101928902
There are no transcripts associated with this gene.
- Gene
- SLC52A2
- Gene Name
- solute carrier family 52 member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001253815.1 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
NP_001240744.1 |
NM_001253816.1 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
NP_001240745.1 |
NM_024531.4 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
NP_078807.1 |
XM_006716658.2 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
XP_006716721.1 |
XM_006716659.2 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
XP_006716722.1 |
XM_006716660.2 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
XP_006716723.1 |
XM_011517300.2 |
1218 |
Silent Mutation |
CCA,CCG |
P131P |
XP_011515602.1 |
XM_017013819.1 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
XP_016869308.1 |
XM_017013820.1 |
1218 |
Silent Mutation |
CCA,CCG |
P219P |
XP_016869309.1 |
XM_017013821.1 |
1218 |
Silent Mutation |
CCA,CCG |
P55P |
XP_016869310.1 |
XM_017013822.1 |
1218 |
Silent Mutation |
CCA,CCG |
P55P |
XP_016869311.1 |
- Gene
- TMEM249
- Gene Name
- transmembrane protein 249
There are no transcripts associated with this gene.
View Full Product Details