Product Details
- SNP ID
-
rs142109502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10898144 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTAATGAGGGGCCCTTCTGGGAG[A/G]TAAGGACGCCCCAACGGGGTAGGGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XKR6
PubMed Links
Gene Details
- Gene
- XKR6
- Gene Name
- XK related 6
View Full Product Details