Product Details
- SNP ID
-
rs142219728
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:52628062 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACCTGGTTTGAGATCCAGGGCAG[A/G]TAGAGACTCTGAATGTAGAAAATAT
- Phenotype
-
MIM: 606837
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RB1CC1
PubMed Links
Gene Details
- Gene
- RB1CC1
- Gene Name
- RB1 inducible coiled-coil 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001083617.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1536S |
NP_001077086.1 |
NM_014781.4 |
5164 |
Missense Mutation |
CCT,TCT |
P1536S |
NP_055596.3 |
XM_011517643.2 |
5164 |
Missense Mutation |
CCT,TCT |
P1536S |
XP_011515945.1 |
XM_011517644.2 |
5164 |
Missense Mutation |
CCT,TCT |
P1536S |
XP_011515946.1 |
XM_011517645.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1536S |
XP_011515947.1 |
XM_011517646.2 |
5164 |
Missense Mutation |
CCT,TCT |
P1484S |
XP_011515948.1 |
XM_011517647.2 |
5164 |
Intron |
|
|
XP_011515949.1 |
XM_011517649.2 |
5164 |
Intron |
|
|
XP_011515951.1 |
XM_017014103.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1543S |
XP_016869592.1 |
XM_017014104.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1543S |
XP_016869593.1 |
XM_017014105.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1543S |
XP_016869594.1 |
XM_017014106.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1543S |
XP_016869595.1 |
XM_017014107.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1536S |
XP_016869596.1 |
XM_017014108.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1543S |
XP_016869597.1 |
XM_017014109.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1491S |
XP_016869598.1 |
XM_017014110.1 |
5164 |
Missense Mutation |
CCT,TCT |
P1000S |
XP_016869599.1 |
XM_017014111.1 |
5164 |
Missense Mutation |
CCT,TCT |
P993S |
XP_016869600.1 |
XM_017014112.1 |
5164 |
Missense Mutation |
CCT,TCT |
P993S |
XP_016869601.1 |
View Full Product Details