Product Details

SNP ID
rs142879103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144360026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAGTGCCCGCCAGCCCCCATCAA[C/T]GGCACCCCTGGCCCCCCGCTCGACT
Phenotype
MIM: 609076 MIM: 607882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL6 PubMed Links

Gene Details

Gene
FBXL6
Gene Name
F-box and leucine rich repeat protein 6
There are no transcripts associated with this gene.

Gene
LOC101928902
Gene Name
uncharacterized LOC101928902
There are no transcripts associated with this gene.

Gene
SLC52A2
Gene Name
solute carrier family 52 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253815.1 1095 Silent Mutation AAC,AAT N178N NP_001240744.1
NM_001253816.1 1095 Silent Mutation AAC,AAT N178N NP_001240745.1
NM_024531.4 1095 Silent Mutation AAC,AAT N178N NP_078807.1
XM_006716658.2 1095 Silent Mutation AAC,AAT N178N XP_006716721.1
XM_006716659.2 1095 Silent Mutation AAC,AAT N178N XP_006716722.1
XM_006716660.2 1095 Silent Mutation AAC,AAT N178N XP_006716723.1
XM_011517300.2 1095 Silent Mutation AAC,AAT N90N XP_011515602.1
XM_017013819.1 1095 Silent Mutation AAC,AAT N178N XP_016869308.1
XM_017013820.1 1095 Silent Mutation AAC,AAT N178N XP_016869309.1
XM_017013821.1 1095 Intron XP_016869310.1
XM_017013822.1 1095 Intron XP_016869311.1
Gene
TMEM249
Gene Name
transmembrane protein 249
There are no transcripts associated with this gene.

View Full Product Details