Product Details
- SNP ID
-
rs143221092
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:124553169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCACTTGGGGGTTCCCGTTCCTG[A/G]TCTTCAGCTTCACTTTCTGGAATTG
- Phenotype
-
MIM: 608486
MIM: 601445
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MTSS1
PubMed Links
Gene Details
- Gene
- MTSS1
- Gene Name
- metastasis suppressor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282971.1 |
2193 |
Silent Mutation |
GAC,GAT |
D701D |
NP_001269900.1 |
NM_001282974.1 |
2193 |
Silent Mutation |
GAC,GAT |
D672D |
NP_001269903.1 |
NM_014751.5 |
2193 |
Silent Mutation |
GAC,GAT |
D697D |
NP_055566.3 |
XM_005251111.1 |
2193 |
Silent Mutation |
GAC,GAT |
D810D |
XP_005251168.1 |
XM_005251113.1 |
2193 |
Silent Mutation |
GAC,GAT |
D745D |
XP_005251170.1 |
XM_005251118.1 |
2193 |
Silent Mutation |
GAC,GAT |
D636D |
XP_005251175.1 |
XM_006716700.1 |
2193 |
Silent Mutation |
GAC,GAT |
D809D |
XP_006716763.1 |
XM_006716701.1 |
2193 |
Silent Mutation |
GAC,GAT |
D806D |
XP_006716764.1 |
XM_006716702.1 |
2193 |
Silent Mutation |
GAC,GAT |
D793D |
XP_006716765.1 |
XM_006716703.1 |
2193 |
Silent Mutation |
GAC,GAT |
D728D |
XP_006716766.1 |
XM_006716704.1 |
2193 |
Silent Mutation |
GAC,GAT |
D700D |
XP_006716767.1 |
XM_006716705.1 |
2193 |
Silent Mutation |
GAC,GAT |
D684D |
XP_006716768.1 |
XM_006716706.1 |
2193 |
Silent Mutation |
GAC,GAT |
D619D |
XP_006716769.1 |
XM_011517403.1 |
2193 |
Silent Mutation |
GAC,GAT |
D743D |
XP_011515705.1 |
XM_011517404.2 |
2193 |
Silent Mutation |
GAC,GAT |
D729D |
XP_011515706.1 |
XM_011517407.1 |
2193 |
Silent Mutation |
GAC,GAT |
D452D |
XP_011515709.1 |
XM_017014086.1 |
2193 |
Silent Mutation |
GAC,GAT |
D805D |
XP_016869575.1 |
XM_017014087.1 |
2193 |
Silent Mutation |
GAC,GAT |
D785D |
XP_016869576.1 |
XM_017014088.1 |
2193 |
Silent Mutation |
GAC,GAT |
D781D |
XP_016869577.1 |
XM_017014089.1 |
2193 |
Silent Mutation |
GAC,GAT |
D780D |
XP_016869578.1 |
XM_017014090.1 |
2193 |
Silent Mutation |
GAC,GAT |
D755D |
XP_016869579.1 |
XM_017014091.1 |
2193 |
Silent Mutation |
GAC,GAT |
D747D |
XP_016869580.1 |
XM_017014092.1 |
2193 |
Silent Mutation |
GAC,GAT |
D741D |
XP_016869581.1 |
XM_017014093.1 |
2193 |
Silent Mutation |
GAC,GAT |
D696D |
XP_016869582.1 |
XM_017014094.1 |
2193 |
Silent Mutation |
GAC,GAT |
D680D |
XP_016869583.1 |
XM_017014095.1 |
2193 |
Silent Mutation |
GAC,GAT |
D676D |
XP_016869584.1 |
XM_017014096.1 |
2193 |
Silent Mutation |
GAC,GAT |
D675D |
XP_016869585.1 |
XM_017014097.1 |
2193 |
Silent Mutation |
GAC,GAT |
D671D |
XP_016869586.1 |
XM_017014098.1 |
2193 |
Silent Mutation |
GAC,GAT |
D632D |
XP_016869587.1 |
XM_017014099.1 |
2193 |
Silent Mutation |
GAC,GAT |
D631D |
XP_016869588.1 |
XM_017014100.1 |
2193 |
Silent Mutation |
GAC,GAT |
D615D |
XP_016869589.1 |
XM_017014101.1 |
2193 |
Silent Mutation |
GAC,GAT |
D591D |
XP_016869590.1 |
- Gene
- NDUFB9
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B9
There are no transcripts associated with this gene.
View Full Product Details