Product Details

SNP ID
rs143671228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52940220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGCTGCGGCAGTGGCCCTTC[C/G]GGGAGCTCATGTGCAAGCTCATCGT
Phenotype
MIM: 600730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NPBWR1 PubMed Links

Gene Details

Gene
NPBWR1
Gene Name
neuropeptides B/W receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005285.3 313 Missense Mutation CGG,GGG R105G NP_005276.2

View Full Product Details