Product Details

SNP ID
rs143819461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144413846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGTGGCTGTGGCCCCCGTGGCT[A/G]TGGCTGCTGTGGCCGCAGGGCCCGT
Phenotype
MIM: 606027 MIM: 607059 MIM: 611952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPSF1 PubMed Links

Gene Details

Gene
CPSF1
Gene Name
cleavage and polyadenylation specific factor 1
There are no transcripts associated with this gene.

Gene
SLC39A4
Gene Name
solute carrier family 39 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280557.1 1408 Intron NP_001267486.1
NM_017767.2 1408 Silent Mutation CAC,CAT H416H NP_060237.2
NM_130849.3 1408 Silent Mutation CAC,CAT H441H NP_570901.2
XM_006716599.1 1408 Silent Mutation CAC,CAT H441H XP_006716662.1
Gene
VPS28
Gene Name
VPS28, ESCRT-I subunit
There are no transcripts associated with this gene.

View Full Product Details