Product Details

SNP ID
rs144768772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144881674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGCCTTCCCGCAGCTATTGCA[C/T]TCATATGGCTTCTCGCCCGTGTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF250 PubMed Links

Gene Details

Gene
ZNF250
Gene Name
zinc finger protein 250
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109689.3 1641 Silent Mutation GAA,GAG E503E NP_001103159.1
NM_021061.4 1641 Silent Mutation GAA,GAG E508E NP_066405.1
XM_005272327.1 1641 Silent Mutation GAA,GAG E503E XP_005272384.1
XM_005272328.3 1641 Silent Mutation GAA,GAG E503E XP_005272385.1
XM_006716612.3 1641 Silent Mutation GAA,GAG E508E XP_006716675.1
XM_006716613.3 1641 Silent Mutation GAA,GAG E508E XP_006716676.1
XM_011517209.1 1641 Silent Mutation GAA,GAG E503E XP_011515511.1

View Full Product Details