Product Details

SNP ID
hCV174186852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142702951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGGCCTGGCCCTCGTCCTCCC[C/G]ACTCCCCCTTGGTGCCCCAGTTGAC
Phenotype
MIM: 615093
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
LY6K
Gene Name
lymphocyte antigen 6 complex, locus K
There are no transcripts associated with this gene.

View Full Product Details