Product Details

SNP ID
rs145615792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81703007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTATTGTCATTTTTAAGCCTGG[C/G]TAGAGAAGCGGCAAAGTCTATGGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC10A5 PubMed Links

Gene Details

Gene
SLC10A5
Gene Name
solute carrier family 10 member 5
There are no transcripts associated with this gene.

Gene
ZFAND1
Gene Name
zinc finger AN1-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170796.1 600 Missense Mutation CCC,GCC P193A NP_001164267.1
NM_001170797.1 600 Missense Mutation CCC,GCC P200A NP_001164268.1
NM_024699.2 600 Missense Mutation CCC,GCC P200A NP_078975.2

View Full Product Details