Product Details
- SNP ID
-
rs145655091
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:61503410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTGGTGTCAGTTCCGGATGCCA[C/T]ACATCCACGATGAATATCAGCCGGA
- Phenotype
-
MIM: 600582
MIM: 611292
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ASPH
PubMed Links
Gene Details
- Gene
- ASPH
- Gene Name
- aspartate beta-hydroxylase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164750.1 |
2278 |
Silent Mutation |
GTA,GTG |
V713V |
NP_001158222.1 |
NM_001164751.1 |
2278 |
Intron |
|
|
NP_001158223.1 |
NM_001164752.1 |
2278 |
Intron |
|
|
NP_001158224.1 |
NM_001164753.1 |
2278 |
Intron |
|
|
NP_001158225.1 |
NM_001164754.1 |
2278 |
Intron |
|
|
NP_001158226.1 |
NM_001164755.1 |
2278 |
Intron |
|
|
NP_001158227.1 |
NM_001164756.1 |
2278 |
Intron |
|
|
NP_001158228.1 |
NM_004318.3 |
2278 |
Silent Mutation |
GTA,GTG |
V742V |
NP_004309.2 |
NM_020164.4 |
2278 |
Intron |
|
|
NP_064549.1 |
NM_032466.3 |
2278 |
Intron |
|
|
NP_115855.1 |
NM_032467.3 |
2278 |
Intron |
|
|
NP_115856.1 |
NM_032468.4 |
2278 |
Intron |
|
|
NP_115857.1 |
XM_005251235.2 |
2278 |
Silent Mutation |
GTA,GTG |
V757V |
XP_005251292.1 |
XM_005251236.2 |
2278 |
Silent Mutation |
GTA,GTG |
V756V |
XP_005251293.1 |
XM_005251238.2 |
2278 |
Silent Mutation |
GTA,GTG |
V742V |
XP_005251295.1 |
XM_005251239.2 |
2278 |
Silent Mutation |
GTA,GTG |
V738V |
XP_005251296.1 |
XM_005251240.1 |
2278 |
Silent Mutation |
GTA,GTG |
V728V |
XP_005251297.1 |
XM_005251242.2 |
2278 |
Silent Mutation |
GTA,GTG |
V723V |
XP_005251299.1 |
XM_005251243.2 |
2278 |
Silent Mutation |
GTA,GTG |
V714V |
XP_005251300.1 |
XM_005251244.1 |
2278 |
Silent Mutation |
GTA,GTG |
V708V |
XP_005251301.1 |
XM_005251246.2 |
2278 |
Silent Mutation |
GTA,GTG |
V699V |
XP_005251303.1 |
XM_005251247.2 |
2278 |
Silent Mutation |
GTA,GTG |
V695V |
XP_005251304.1 |
XM_005251248.1 |
2278 |
Silent Mutation |
GTA,GTG |
V694V |
XP_005251305.1 |
XM_005251250.2 |
2278 |
Silent Mutation |
GTA,GTG |
V680V |
XP_005251307.1 |
XM_017013419.1 |
2278 |
Silent Mutation |
GTA,GTG |
V784V |
XP_016868908.1 |
XM_017013420.1 |
2278 |
Silent Mutation |
GTA,GTG |
V783V |
XP_016868909.1 |
XM_017013421.1 |
2278 |
Silent Mutation |
GTA,GTG |
V770V |
XP_016868910.1 |
XM_017013422.1 |
2278 |
Silent Mutation |
GTA,GTG |
V769V |
XP_016868911.1 |
XM_017013423.1 |
2278 |
Silent Mutation |
GTA,GTG |
V769V |
XP_016868912.1 |
XM_017013424.1 |
2278 |
Silent Mutation |
GTA,GTG |
V768V |
XP_016868913.1 |
XM_017013425.1 |
2278 |
Silent Mutation |
GTA,GTG |
V765V |
XP_016868914.1 |
XM_017013426.1 |
2278 |
Silent Mutation |
GTA,GTG |
V755V |
XP_016868915.1 |
XM_017013427.1 |
2278 |
Silent Mutation |
GTA,GTG |
V754V |
XP_016868916.1 |
XM_017013428.1 |
2278 |
Silent Mutation |
GTA,GTG |
V750V |
XP_016868917.1 |
XM_017013429.1 |
2278 |
Silent Mutation |
GTA,GTG |
V741V |
XP_016868918.1 |
XM_017013430.1 |
2278 |
Silent Mutation |
GTA,GTG |
V741V |
XP_016868919.1 |
XM_017013431.1 |
2278 |
Silent Mutation |
GTA,GTG |
V740V |
XP_016868920.1 |
XM_017013432.1 |
2278 |
Silent Mutation |
GTA,GTG |
V740V |
XP_016868921.1 |
XM_017013433.1 |
2278 |
Silent Mutation |
GTA,GTG |
V739V |
XP_016868922.1 |
XM_017013434.1 |
2278 |
Silent Mutation |
GTA,GTG |
V737V |
XP_016868923.1 |
XM_017013435.1 |
2278 |
Silent Mutation |
GTA,GTG |
V735V |
XP_016868924.1 |
XM_017013436.1 |
2278 |
Silent Mutation |
GTA,GTG |
V728V |
XP_016868925.1 |
XM_017013437.1 |
2278 |
Silent Mutation |
GTA,GTG |
V726V |
XP_016868926.1 |
XM_017013438.1 |
2278 |
Silent Mutation |
GTA,GTG |
V713V |
XP_016868927.1 |
XM_017013439.1 |
2278 |
Silent Mutation |
GTA,GTG |
V712V |
XP_016868928.1 |
XM_017013440.1 |
2278 |
Silent Mutation |
GTA,GTG |
V712V |
XP_016868929.1 |
XM_017013441.1 |
2278 |
Silent Mutation |
GTA,GTG |
V698V |
XP_016868930.1 |
XM_017013442.1 |
2278 |
Silent Mutation |
GTA,GTG |
V694V |
XP_016868931.1 |
XM_017013443.1 |
2278 |
Silent Mutation |
GTA,GTG |
V679V |
XP_016868932.1 |
XM_017013444.1 |
2278 |
Silent Mutation |
GTA,GTG |
V670V |
XP_016868933.1 |
XM_017013445.1 |
2278 |
Intron |
|
|
XP_016868934.1 |
XM_017013446.1 |
2278 |
Intron |
|
|
XP_016868935.1 |
XM_017013447.1 |
2278 |
Intron |
|
|
XP_016868936.1 |
- Gene
- CLVS1
- Gene Name
- clavesin 1
There are no transcripts associated with this gene.
View Full Product Details