Product Details

SNP ID
rs145660272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142741253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGGAGCAGGAGCGGGTCACCA[C/T]GGGGCTCTGGTTGAAGGGGTACTCT
Phenotype
MIM: 606119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYPD2 PubMed Links

Gene Details

Gene
LYPD2
Gene Name
LY6/PLAUR domain containing 2
There are no transcripts associated with this gene.

Gene
SLURP1
Gene Name
secreted LY6/PLAUR domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020427.2 228 Missense Mutation ATG,GTG M68V NP_065160.1
Gene
THEM6
Gene Name
thioesterase superfamily member 6
There are no transcripts associated with this gene.

View Full Product Details