Product Details

SNP ID
rs145823107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58806559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGGGCAATTCTGTTATGTCGT[A/G]TGAAGAAATAAAGTTCTTAAATATA
Phenotype
MIM: 606863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOX PubMed Links

Gene Details

Gene
TOX
Gene Name
thymocyte selection associated high mobility group box
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014729.2 3009 UTR 3 NP_055544.1
XM_017014085.1 3009 UTR 3 XP_016869574.1

View Full Product Details