Product Details

SNP ID
rs145996576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38107164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGACACATTTGGCATAGATAC[A/C]TCGTGAGTACTTTTCATAGTTTTTG
Phenotype
MIM: 604782
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASH2L PubMed Links

Gene Details

Gene
ASH2L
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105214.2 558 Silent Mutation ACA,ACC T39T NP_001098684.1
NM_001261832.1 558 Silent Mutation ACA,ACC T39T NP_001248761.1
NM_001282272.1 558 Intron NP_001269201.1
NM_004674.4 558 Silent Mutation ACA,ACC T133T NP_004665.2
XM_005273682.1 558 Silent Mutation ACA,ACC T133T XP_005273739.1
XM_005273683.1 558 Silent Mutation ACA,ACC T39T XP_005273740.1
XM_006716412.1 558 Silent Mutation ACA,ACC T133T XP_006716475.1
XM_006716413.2 558 Intron XP_006716476.1

View Full Product Details