Product Details

SNP ID
rs146036612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:112223210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTGCATCCTGCCAGTAGAGTA[C/G]CATGTTGAGAAAATTGTATGAATTT
Phenotype
MIM: 608399
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSMD3 PubMed Links

Gene Details

Gene
CSMD3
Gene Name
CUB and Sushi multiple domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052900.2 13252 UTR 3 NP_443132.3
NM_198123.1 13252 UTR 3 NP_937756.1
NM_198124.1 13252 UTR 3 NP_937757.1
XM_011516815.2 13252 UTR 3 XP_011515117.1
XM_011516816.2 13252 UTR 3 XP_011515118.1
XM_017013008.1 13252 UTR 3 XP_016868497.1
XM_017013009.1 13252 UTR 3 XP_016868498.1
XM_017013010.1 13252 UTR 3 XP_016868499.1
XM_017013011.1 13252 UTR 3 XP_016868500.1
XM_017013012.1 13252 UTR 3 XP_016868501.1

View Full Product Details